Learn More
Waters Corp Waters Silica Column, 80Å, 5 μm, 2 mm × 100 mm
Catalog No. 50784296 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Spherisorb silica chemistry is a general purpose unbonded silica substrate designed for normal phase applications. It is a highly specialized HPLC column designed for narrowbore instrumentation. With a 2 mm inner diameter and a 10 cm length, it is suited for high-resolution separations, delivering exceptional peak shape and efficiency. The column's 5 μm particle size and end-capped design reduce column bleed and improve performance. It is compatible with a wide range of solvents and can withstand high temperatures and pressures, making it a versatile and reliable choice for demanding HPLC applications.
  • High-resolution separations with enhanced peak shape and resolution.
  • Reduced column bleed with 5 μm particle size and end-capping.
  • Versatile solvent compatibility for water, acetonitrile, methanol, and tetrahydrofuran.
  • Robust operation, withstanding 45°C and 6000 psi (415 Bar).
Catalog No. 50-784-296 Supplier Waters Corp Supplier No. PSS830122
The Spherisorb silica chemistry is a general purpose unbonded silica substrate designed for normal phase applications. It is a highly specialized HPLC column designed for narrowbore instrumentation. With a 2 mm inner diameter and a 10 cm length, it is suited for high-resolution separations, delivering exceptional peak shape and efficiency. The column's 5 μm particle size and end-capped design reduce column bleed and improve performance. It is compatible with a wide range of solvents and can withstand high temperatures and pressures, making it a versatile and reliable choice for demanding HPLC applications.
  • High-resolution separations with enhanced peak shape and resolution.
  • Reduced column bleed with 5 μm particle size and end-capping.
  • Versatile solvent compatibility for water, acetonitrile, methanol, and tetrahydrofuran.
  • Robust operation, withstanding 45°C and 6000 psi (415 Bar).