Learn More
Waters Corp Silica Column, 3 μm, 4.6 mm ID, 150 mm
Catalog No. 50784399 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column features Spherisorb silica chemistry, serving as a general-purpose unbonded silica substrate for normal phase applications. It is manufactured with high-quality bonded phases, providing a wide array of separation selectivities, and incorporates industry-standard Parker-style column end fittings.
  • General-purpose unbonded silica substrate
  • Designed for normal phase applications
  • High-quality bonded phases for diverse separation selectivities
  • Industry-standard Parker-style column end fittings
  • Produced at cGMP-compliant facilities
  • ISO 9002 certified
  • FDA-registered as a Class 1 medical device manufacturer
Catalog No. 50-784-399 Supplier Waters Corp Supplier No. PSS832013
This column features Spherisorb silica chemistry, serving as a general-purpose unbonded silica substrate for normal phase applications. It is manufactured with high-quality bonded phases, providing a wide array of separation selectivities, and incorporates industry-standard Parker-style column end fittings.
  • General-purpose unbonded silica substrate
  • Designed for normal phase applications
  • High-quality bonded phases for diverse separation selectivities
  • Industry-standard Parker-style column end fittings
  • Produced at cGMP-compliant facilities
  • ISO 9002 certified
  • FDA-registered as a Class 1 medical device manufacturer