Learn More
Waters Corp Spherisorb Silica Column, 80Å, 5μm, 50 mm Length
Catalog No. 50784628 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Spherisorb Silica Column is designed for chromatography applications. It features specific dimensions and particle characteristics suitable for analytical separations.
  • Features specific dimensions, including a 2.0 mm ID and 50 mm length.
  • Equipped with a 5μm particle size.
  • Offers an 80Å pore size.
  • Classified as USP L3.
Catalog No. 50-784-628 Supplier Waters Corp Supplier No. PSS839840
This Spherisorb Silica Column is designed for chromatography applications. It features specific dimensions and particle characteristics suitable for analytical separations.
  • Features specific dimensions, including a 2.0 mm ID and 50 mm length.
  • Equipped with a 5μm particle size.
  • Offers an 80Å pore size.
  • Classified as USP L3.