Learn More
Waters Corp Spherisorb Silica Column, 80Å, 3 µm, 2 mm X 150 mm, 1/pk
Catalog No. 50784401 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Spherisorb silica chemistry is general purpose unbonded silica substrate designed for normal phase applications.
Catalog No. 50-784-401 Supplier Waters Corp Supplier No. PSS832023
The Spherisorb silica chemistry is general purpose unbonded silica substrate designed for normal phase applications.