Learn More
Waters Corp Spherisorb Silica Guard Cartridge, 80Å, 5 µm, 4.6 mm X 30 mm, 3/pk
Catalog No. 50784528 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Spherisorb silica chemistry is general purpose unbonded silica substrate designed for normal phase applications.
Catalog No. 50-784-528 Supplier Waters Corp Supplier No. PSS839451
The Spherisorb silica chemistry is general purpose unbonded silica substrate designed for normal phase applications.