Learn More
Waters Corp Waters Corporation Guard Cartridge, Silica, 80Å, 5 μm, 4.6 mm × 10 mm
Catalog No. 50784277 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Spherisorb guard cartridge, featuring silica chemistry, protects HPLC columns from particulate matter and organic solvents in normal phase applications. Built with 80Å silica and a 5 μm particle size, it enhances system longevity and performance through optimal filtration and stability across diverse solvents and temperatures.
  • Protects HPLC columns from particulate matter and organic solvents.
  • Utilizes 80Å silica and 5 μm particle size for optimal filtration.
  • Stable across a wide range of solvents and temperatures.
  • Easy to install, enhancing HPLC system longevity and performance.
  • Reduces need for costly column replacements.
Catalog No. 50-784-277 Supplier Waters Corp Supplier No. PSS830051
This Spherisorb guard cartridge, featuring silica chemistry, protects HPLC columns from particulate matter and organic solvents in normal phase applications. Built with 80Å silica and a 5 μm particle size, it enhances system longevity and performance through optimal filtration and stability across diverse solvents and temperatures.
  • Protects HPLC columns from particulate matter and organic solvents.
  • Utilizes 80Å silica and 5 μm particle size for optimal filtration.
  • Stable across a wide range of solvents and temperatures.
  • Easy to install, enhancing HPLC system longevity and performance.
  • Reduces need for costly column replacements.