Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Qualification Standards Test Kit, TQD/Q-Premier/XE/Q-Micro, 10 Component, Mass Spectrometry

Catalog No. 50792778 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The TQD/Q-Premier/XE/Q-Micro Qualification Standards Test Kit is part of the MS Triple Quadrupole Standards collection series. This kit is designed to enable scientists to assess the performance or calibrate the triple quadrupole detector. It facilitates mass scale calibration, resolution, and sensitivity testing across various ionization modes. Using this kit ensures optimal MS performance, providing confidence in assay results.

  • Mass spectrometry standards kit for system assessment.
  • 10 Component kit including various solutions and solids for specific tests.
  • Separation mode: Reversed phase.
  • Detection method: MS.
  • Formulation: Liquid, solid.
  • Storage condition: Room temperature.
  • System type: MS.
  • Application: System performance.
  • Includes Certificate of Analysis and Solution Prep Document.

Catalog No. 50-792-778 Supplier Waters Corp Supplier No. 700002646
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TQD/Q-Premier/XE/Q-Micro Qualification Standards Test Kit is part of the MS Triple Quadrupole Standards collection series. This kit is designed to enable scientists to assess the performance or calibrate the triple quadrupole detector. It facilitates mass scale calibration, resolution, and sensitivity testing across various ionization modes. Using this kit ensures optimal MS performance, providing confidence in assay results.

  • Mass spectrometry standards kit for system assessment.
  • 10 Component kit including various solutions and solids for specific tests.
  • Separation mode: Reversed phase.
  • Detection method: MS.
  • Formulation: Liquid, solid.
  • Storage condition: Room temperature.
  • System type: MS.
  • Application: System performance.
  • Includes Certificate of Analysis and Solution Prep Document.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.