Learn More
Optimize Technologies Waters Type Union w/ Nuts & Ferrules for 1/16" OD Tubing
SDP
Supplier:  Optimize Technologies 392401088
Encompass_Preferred
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi. We realize that your time is valuable and should not be wasted worrying about fittings. For this reason, all Optimize fittings are precision machined in the USA for optimal quality and thread consistency. The machining process is the best way to ensure tight manufacturing tolerances and minimal product variance. This product is equivalent to OEM part number WAT097332.
Catalog No. 50-114-117