Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Watershed Tour
SDPSCI_ED

Catalog No. S65399 Shop All LaMotte Company Products
Change view
Click to view available options
Product Type:
Watershed Tour Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S65399 Watershed Tour Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65399 Supplier LaMotte™ Supplier No. 5419

missing translation for 'validMainframeMsg'

Includes

Lecture materials, illustrated handouts, teacher tips, test procedures, TesTabs™ reagents, data sheets and games

Kit guides the class through a week-long series of activities focused on stream and river ecosystems.

Designed for teachers who are unable to visit a watershed with their students

  • Teaches students about stream ecology, water quality issues and their own connection to a watershed
  • Enables students to perform simulated tests on four sites along a river continuum — from the headwaters to the mouth of the river as it enters the estuary — to study how the river changes and how human activities can influence water quality

Order Info

Grades 4 to 8. Class size: 30 students working in groups.

Specifications

Grade 4 to 8
Class Size 1 Class
Product Type Watershed Tour Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.