Learn More
Across International Ai WaterVac 0.7 cfm 2-Head Water Aspirator Vacuum Pump
SDP
Catalog No. NC1971796
Encompass_Preferred
Ai WaterVac 0.7 cfm is an oil-free, portable, chemical resistant, 2-head recirculating water aspirator pump, with simplified maintenance, and no frequent and costly oil change. All major parts are made of PTFE, these economical vacuum pumps are perfect for tough lab applications.
Catalog No. NC1971796 Supplier Across International Supplier No. WV07.110
Ai WaterVac 0.7 cfm is an oil-free, portable, chemical resistant, 2-head recirculating water aspirator pump, with simplified maintenance, and no frequent and costly oil change. All major parts are made of PTFE, these economical vacuum pumps are perfect for tough lab applications.