Learn More
CTL Scientific Supply Corp WATOR, 50 PIECES
SDP
Catalog No. 501173284 Shop All CTL Scientific Supply Products
Encompass_Preferred
WATOR - Indicator paper for the determination of water in butter according to DIN 10 311. Box of 50 pieces (each piece 78 x 40 mm). On contact with water WATOR develops dark-blue spots, the size and number of which is correlated to the distribution of the water in the butter.
Catalog No. 50-117-3284 Supplier CTL Scientific Supply Corp Supplier No. 90610
WATOR - Indicator paper for the determination of water in butter according to DIN 10 311. Box of 50 pieces (each piece 78 x 40 mm). On contact with water WATOR develops dark-blue spots, the size and number of which is correlated to the distribution of the water in the butter.