Learn More
Alkali Scientific CellPro™ Molecular Biological Grade Water, 20 Liters
SDP
Catalog No. 502335826
Encompass_Preferred
CellPro™ Molecular Biological Grade Water is suitable for use in molecular biology applications which demand a high quality of water and assurance that the water is free from DNase and RNase contamination. Molecular Grade Water can be used for making reagents and rinsing glass/plastic wares for DNA and RNA research.
Catalog No. 50-233-5826 Supplier Alkali Scientific Supplier No. DC20L
CellPro™ Molecular Biological Grade Water is suitable for use in molecular biology applications which demand a high quality of water and assurance that the water is free from DNase and RNase contamination. Molecular Grade Water can be used for making reagents and rinsing glass/plastic wares for DNA and RNA research.