Learn More
Watson-Marlow 313 Rapid-Load Pumphead Extension
SDP
Catalog No. 14283106 Shop All Watson-Marlow Bredel Products
Change view
Click to view available options
For Use With:
Metering and transfer pump
Catalog No. For Use With
14-283-106 Metering and transfer pump
1 options
Catalog No. 14-283-106 Supplier Watson-Marlow Supplier No. 033.3431.000
Add to Cart
Add to Cart
Offers simple, error-free tube loading
  • Pumphead accepts seven tube sizes to meet any flow requirement
  • Three nylatron rollers
  • Tube wall of 1.6mm
  • With adjustable tube clamps
TRUSTED_SUSTAINABILITY
Specifications
For Use With Metering and transfer pump
Holds 3
Includes Adjustable tube clamps
Roller Material Nylatron
Type Extension Pump Head