Learn More
WATSON MARLOW INC Watson-Marlow Peristaltic Pump System, 323Du/D Drive, 313D Three-Roller, 400 rpm, RS232 Control
SDP
Catalog No. NC3989759
Encompass
This 300 series laboratory pump is a countertop pumping solution offering manual, remote, analogue, or RS232 control. It features a chemically resistant ABS top, IP31 protection, a membrane keypad for control, and a digital LCD to display speed.
  • Manual, remote, and analogue auto-control or RS232 control commands
  • Enhanced digital manual speed control up to 133:1 with 3 to 400 rpm range
  • Keypad lock prevents tampering or accidental changes
  • MemoDose facility for accurate single-shot dosing
  • Digital TTL inputs for run/stop and direction control
  • Instantly reversible operation
  • Two-year warranty
  • 1 rpm scrolling increment for precise adjustments
  • Auto-restart capability
  • IP31 wipe-down case enclosure
  • RS232 serial link for digital communication
Catalog No. NC3989759 Supplier WATSON MARLOW INC Supplier No. 030.3154.3DA
This 300 series laboratory pump is a countertop pumping solution offering manual, remote, analogue, or RS232 control. It features a chemically resistant ABS top, IP31 protection, a membrane keypad for control, and a digital LCD to display speed.
  • Manual, remote, and analogue auto-control or RS232 control commands
  • Enhanced digital manual speed control up to 133:1 with 3 to 400 rpm range
  • Keypad lock prevents tampering or accidental changes
  • MemoDose facility for accurate single-shot dosing
  • Digital TTL inputs for run/stop and direction control
  • Instantly reversible operation
  • Two-year warranty
  • 1 rpm scrolling increment for precise adjustments
  • Auto-restart capability
  • IP31 wipe-down case enclosure
  • RS232 serial link for digital communication