Learn More
Cayman Chemical WaTx36-68scorpIontrIf 10mg
SDP
Catalog No. 502918087
Encompass_Preferred
A cell-penetrating peptide and TRPA1 activator; selectively induces currents in HEK293T cells expressing human TRPA1 (EC50 = 15 nM) over cells expressing human TRPM8 or TRPV1 at 100 nM or rat Kv1.2, Kv2.1, Kv3.1, or Kv4.3 at 1,000 nM; induces nocifensive behavior in wild-type but not Trpa1-/- mice
Catalog No. 50-291-8087 Supplier Cayman Chemical Supplier No. 3749510 MG
A cell-penetrating peptide and TRPA1 activator; selectively induces currents in HEK293T cells expressing human TRPA1 (EC50 = 15 nM) over cells expressing human TRPM8 or TRPV1 at 100 nM or rat Kv1.2, Kv2.1, Kv3.1, or Kv4.3 at 1,000 nM; induces nocifensive behavior in wild-type but not Trpa1-/- mice