Learn More
Sklar Instruments Waugh Dressing Forceps, Sklar, 8 in., Straight, Cross-Serrated, Stainless Steel.
SDP
Catalog No. 501437429 Shop All Sklar Instruments Products
Encompass_Preferred
Waugh Dressing Forceps, Sklar, 8 in., Straight, Cross-Serrated, Stainless Steel. Sklar Waugh Forceps are thumb forceps used for both dissection and holding tissue or dressings. This product is straight with cross-serrated tips and a length of 8 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7429 Supplier Sklar Instruments Supplier No. 192189
Waugh Dressing Forceps, Sklar, 8 in., Straight, Cross-Serrated, Stainless Steel. Sklar Waugh Forceps are thumb forceps used for both dissection and holding tissue or dressings. This product is straight with cross-serrated tips and a length of 8 inches.