Learn More
GSC Go Science Crazy Wave Demonstration Kit
SCI_ED
Catalog No. S24655 Shop All GSC Products
Encompass
Encompass_Preferred
Catalog No. S24655 Supplier GSC Go Science Crazy Supplier No. WVDEM02
A mesmerizing demonstration of wave patterns
The pendulum wave demonstration is a cross between science and art. It demonstrates how mass accelerates and decelerates as a response to the force of gravity. Nine 1 in diameter steel pendulums will form wave patterns as they fall in and out of sync with one another. Some minor assembly is required. Instructions and teachers guide included.
Specifications
Product Type Pendulum Wave Demonstration
For Use With (Application) Demonstrates how mass accelerates and decelerates as a response to the force of gravity
Includes Nine 1 in. diameter steel pendulums and frame
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov