Learn More
Wave Generators
SDPSCI_ED
Catalog No. S52082
Change view
Click to view available options
Variable Phase Wave Generator
Catalog No.
S52082 Variable Phase Wave Generator
1 options
Catalog No. S52082 Supplier Science First Supplier No. 15445
Variable or adjustable phase wave generators available to show different wave effects.
Variable Phase, Two-Point Source
  • Phase relationship can be continuously adjusted from 0° to 360° without interrupting the wave pattern
  • Circular waves from a single point can also be generated
  • Mounted inside or outside the tank
Adjustable Phase, Two-Point Source
  • Phase relationship can be adjusted from 0° to 360°
  • Produce both circular and straight waves of various frequencies
  • Mounted inside or outside the ripple tank
Specifications
Type Adjustable Phase Circular and Straight Wave Generator
For Use With (Application) To show different wave effects
Includes Removable wave bar and two cam actuated rods