Learn More
UNITRON Wave LED Magnifier, 45 in. Arm, 5-D Lens and Edge Clamp, White
SDP
Catalog No. 01189333 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Color:
White
Catalog No. Color
01-189-333 White
1 options
Catalog No. 01-189-333 Supplier UNITRON Supplier No. WAL026445
Add to Cart
Add to Cart
Delivers shadow-free illumination and enhanced 3D magnification, ideal for precision and delicate tasks.
  • Dual 6 W dimmable LED modules with 13 W total consumption, step dimming (0-50-100%), 4600 lux at 11 in. focal length, 4000 K CCT, and CRI 80 for clear, consistent lighting.
  • Includes standard edge clamp for secure and stable mounting during use.
  • 5-diopter (2.25X) magnification with 6.75 x 4.5 in. rectangular white crown optical-quality glass lenses for sharp visual clarity.
  • Supplied with power cable and plug, featuring integrated auto shut-off after 4 or 9 hours for energy efficiency and safety.
  • Heavy-duty 45 in. internal-spring, three-pivot K-arm with self-balancing shade and hands-free neck for precise, drift-free positioning.
Specifications
Type Magnifier
Color White