Learn More
BURTON MEDICAL LLC WAVE LED
Catalog No. NC1007870
Encompass_Preferred
4600 lux at 11" (28 cm). 4000 K Color temperature. CRI of 80. 50, 000 hour average LED Life. Standard table edge clamp mount included. Weighted floor stand sold seperately.
Catalog No. NC1007870 Supplier BURTON MEDICAL LLC Supplier No. WML35W45EC120
4600 lux at 11" (28 cm). 4000 K Color temperature. CRI of 80. 50, 000 hour average LED Life. Standard table edge clamp mount included. Weighted floor stand sold seperately.