Learn More
Perkin Elmer US LLC Instrument Check Standard 3, 250 mL
Catalog No. 509043660
Encompass_Preferred
100 ppm K, P, S 20 ppm AS, LA, LI, MN, MO, NA, NI, SC 5% HCL
Catalog No. 50-904-3660 Supplier Perkin Elmer US LLC Supplier No. N0681470
100 ppm K, P, S 20 ppm AS, LA, LI, MN, MO, NA, NI, SC 5% HCL