Learn More
SPEX CERTIPREP LLC 3P WAVECAL2 TUNING SOLUTION 250ML
Catalog No. NC0170359
Encompass_Preferred
(HPS by Zeptometrix) Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn - 50 ug/mL; K - 500 ug/mL in 5% HNO3 Exp Date: 1 Year
Catalog No. NC0170359 Supplier SPEX CERTIPREP LLC 3P Supplier No. WAVECAL2250
(HPS by Zeptometrix) Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn - 50 ug/mL; K - 500 ug/mL in 5% HNO3 Exp Date: 1 Year