Learn More
Malvern Panalytical WAVEchip 4PCH-NTA, 4 channels, NTA-functionalized, disposable microfluidic sensor chip, Compatible with WAVEsystem, Particle Size Compatibility up to 1000 nm
Catalog No. NC3735218
Encompass
WAVEchip 4PCH-NTA, 4 channels, NTA-functionalized, disposable microfluidic sensor chip, Compatible with WAVEsystem, Particle Size Compatibility up to 1000 nm
Catalog No. NC3735218 Supplier Malvern Panalytical Supplier No. 9060009
WAVEchip 4PCH-NTA, 4 channels, NTA-functionalized, disposable microfluidic sensor chip, Compatible with WAVEsystem, Particle Size Compatibility up to 1000 nm