Learn More
Malvern Panalytical WAVECHIP 4PCH-STA, 4-channel biosensor chip, pre-immobilized streptavidin surface, thick hydrogel matrix, no-clog microfluidics
Catalog No. NC3710913
Encompass
WAVECHIP 4PCH-STA, 4-channel biosensor chip, pre-immobilized streptavidin surface, thick hydrogel matrix, no-clog microfluidics
Catalog No. NC3710913 Supplier Malvern Panalytical Supplier No. 9060010
WAVECHIP 4PCH-STA, 4-channel biosensor chip, pre-immobilized streptavidin surface, thick hydrogel matrix, no-clog microfluidics