Learn More
Medical Chemical Corporation Wavicide-01, 1 qt, 12/cs
SDP
Catalog No. NC1540381
Encompass_Preferred
WAVICIDE-01 is a 2.65% glutaraldehyde sterilant / high level disinfectant soaking solution that has been tested and approved for use with endoscopes and stainless steel surgical instruments.
This item is not returnable. View return policy
Catalog No. NC1540381 Supplier Medical Chemical Corporation Supplier No. 011232OZ
WAVICIDE-01 is a 2.65% glutaraldehyde sterilant / high level disinfectant soaking solution that has been tested and approved for use with endoscopes and stainless steel surgical instruments.