Learn More
Med Vet International Bone wax, Sterile, 2.5gm packets, J0912Q, 12 Packets/Box
Catalog No. 5000495520
Encompass_Preferred
This natural wax will control the seepage of blood from cut - cancellous bone, which obscures the visual field. The wax acts as a physical barrier.
Catalog No. 50-004-95520 Supplier Med Vet International Supplier No. JOR912Q
This natural wax will control the seepage of blood from cut - cancellous bone, which obscures the visual field. The wax acts as a physical barrier.