Learn More
Electron Microscopy Sciences Wax Pen 1
SDP
Catalog No. 50949309
Encompass_Preferred
Powered by AA batteries. This pen helps to flatten and separate tissue sections and reduces compression in thin sections. Requires 1 battery.
Catalog No. 50-949-309 Supplier Electron Microscopy Sciences Supplier No. 72678
Powered by AA batteries. This pen helps to flatten and separate tissue sections and reduces compression in thin sections. Requires 1 battery.