Learn More
Electron Microscopy Sciences Wax Pen 2
SDP
Catalog No. 50949319
Encompass_Preferred
Powered by AA batteries. This pen helps to flatten and separate tissue sections and reduces compression in thin sections. Requires 2 battery.
Catalog No. 50-949-319 Supplier Electron Microscopy Sciences Supplier No. 72679
Powered by AA batteries. This pen helps to flatten and separate tissue sections and reduces compression in thin sections. Requires 2 battery.