Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wax Pencils
SCI_ED

Catalog No. S45652B
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S45652B Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S45652B Supplier STAPLES ADVANTAGE Supplier No. 440325

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sold in packs of 12.

Specifications

Product Type Wax Pencil
Color Blue
For Use With (Application) Ideal for writing on china, glass, metal or plastic. Marks on many porous and non-porous surfaces
Quantity 12
Width (Metric) Line 4.7 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.