Learn More
Ted Pella Inc Cavex Regular Wax Sheets, 23 sheets, 500g
SDP
Catalog No. NC9377103
Encompass
Cavex Set Up Regular with a melting point of 58° C (136° F). Cavex has excellent sealing properties and is a well known and worldwide recognized brand for dental applications. Pink in color. Box with 23 sheets 189 x 78 x 1.63mm thick (7-7/6 x 3-1/16 x .064 thick), 500g (1.1 lb.).
Catalog No. NC9377103 Supplier Ted Pella Inc Supplier No. 109
Cavex Set Up Regular with a melting point of 58° C (136° F). Cavex has excellent sealing properties and is a well known and worldwide recognized brand for dental applications. Pink in color. Box with 23 sheets 189 x 78 x 1.63mm thick (7-7/6 x 3-1/16 x .064 thick), 500g (1.1 lb.).