Learn More
Medchemexpress LLC Way-100135 dihydrochloride | 149055-79-8 | 98.1% | 468.46 g/mol | C24H35Cl2N3O2 | 10 MG
SDP
Catalog No. 5000226766
Encompass_Preferred
WAY-100135 dihydrochloride is a selective 5-HT1A receptor antagonist supplied as the dihydrochloride salt for research use. It is typically used in pharmacology studies to probe serotonergic signaling and in models of mood and anxiety disorders.
  • Selective antagonist of presynaptic and postsynaptic 5-HT1A receptors.
  • Reported activity with an IC50 of approximately 34 nM in rat hippocampal tissue.
  • Dihydrochloride salt form enhances solubility for biological assays.
  • High chemical purity suitable for laboratory research.
  • Available in small-format quantities for dose-response and pilot studies.
Catalog No. 50-002-26766 Supplier Medchemexpress LLC Supplier No. HY117575A10MG
WAY-100135 dihydrochloride is a selective 5-HT1A receptor antagonist supplied as the dihydrochloride salt for research use. It is typically used in pharmacology studies to probe serotonergic signaling and in models of mood and anxiety disorders.
  • Selective antagonist of presynaptic and postsynaptic 5-HT1A receptors.
  • Reported activity with an IC50 of approximately 34 nM in rat hippocampal tissue.
  • Dihydrochloride salt form enhances solubility for biological assays.
  • High chemical purity suitable for laboratory research.
  • Available in small-format quantities for dose-response and pilot studies.