Learn More
Medchemexpress LLC Way-100135 dihydrochloride | 149055-79-8 | 98.1% | 468.46 g/mol | C24H35Cl2N3O2 | 5 MG
SDP
Catalog No. 5000226807
Encompass_Preferred
WAY-100135 dihydrochloride is a research-grade small molecule used as a selective antagonist at presynaptic and postsynaptic 5-HT1A receptors. Supplied as the dihydrochloride salt with reported in vitro potency (IC50 ≈ 34 nM) and high purity, it is intended for pharmacology and neuroscience studies.
  • Selective antagonist of 5-HT1A receptors.
  • High purity suitable for analytical and preclinical research.
  • Commonly used for receptor pharmacology and signaling studies.
  • Available in small pack sizes for laboratory experiments.
  • Molecular weight provided for dosing and calculation purposes.
Catalog No. 50-002-26807 Supplier Medchemexpress LLC Supplier No. HY117575A5MG
WAY-100135 dihydrochloride is a research-grade small molecule used as a selective antagonist at presynaptic and postsynaptic 5-HT1A receptors. Supplied as the dihydrochloride salt with reported in vitro potency (IC50 ≈ 34 nM) and high purity, it is intended for pharmacology and neuroscience studies.
  • Selective antagonist of 5-HT1A receptors.
  • High purity suitable for analytical and preclinical research.
  • Commonly used for receptor pharmacology and signaling studies.
  • Available in small pack sizes for laboratory experiments.
  • Molecular weight provided for dosing and calculation purposes.