Learn More
Apexbio Technology LLC WAY-100635 162760-96-5 200mg
SDP
Catalog No. 5000095472
Encompass_Preferred
WAY-100635 (CAS 162760-96-5) is a highly selective antagonist of the serotonin 5-HT1A receptor In vitro studies show that WAY-100635 displaces the 5-HT1A radioligand [3H]8-OH-DPAT from rat hippocampal membranes with a pIC50 of 8 87 and acts as a potent antagonist in functional assays without agonist or partial agonist activity In ex vivo guinea pig ileum assays it effectively blocks 5-carboxamidotryptamine-induced 5-HT1A responses (pA2 9 71 at 0 3 nM) In vivo WAY-100635 counteracts 8-OH-DPAT-induced behavioral and thermoregulatory effects in rodents at subcutaneous doses as low as 0 003 0 01 mg/kg It is widely utilized in neuropharmacological research and neuroimaging as a SPECT/PET ligand to investigate 5-HT1A receptor function in psychiatric and neurological disorders
Catalog No. 50-000-95472 Supplier Apexbio Technology LLC Supplier No. A3933200
WAY-100635 (CAS 162760-96-5) is a highly selective antagonist of the serotonin 5-HT1A receptor In vitro studies show that WAY-100635 displaces the 5-HT1A radioligand [3H]8-OH-DPAT from rat hippocampal membranes with a pIC50 of 8 87 and acts as a potent antagonist in functional assays without agonist or partial agonist activity In ex vivo guinea pig ileum assays it effectively blocks 5-carboxamidotryptamine-induced 5-HT1A responses (pA2 9 71 at 0 3 nM) In vivo WAY-100635 counteracts 8-OH-DPAT-induced behavioral and thermoregulatory effects in rodents at subcutaneous doses as low as 0 003 0 01 mg/kg It is widely utilized in neuropharmacological research and neuroimaging as a SPECT/PET ligand to investigate 5-HT1A receptor function in psychiatric and neurological disorders