Learn More
Medchemexpress LLC WAY-100635 maleate | 1092679-51-0 | MFCD01321056 | 99.6% | 538.64 g·mol⁻1 | C29H38N4O6 | 1 ML
SDP
Catalog No. 5000248624
Encompass_Preferred
WAY-100635 maleate is a potent and selective 5-HT1A receptor antagonist used in pharmacological research. It is supplied as a high-purity solid or as a 10 mM solution in DMSO and is intended for in vitro and in vivo studies requiring serotonin 5-HT1A receptor blockade.
  • Potent, selective 5-HT1A receptor antagonist.
  • Available as 10 mM solution in DMSO or as solid powder.
  • High purity (≈99.6%) suitable for research use.
  • Recommended storage: solid at 4°C; solution at -20°C (short term) or -80°C (long term).
Catalog No. 50-002-48624 Supplier Medchemexpress LLC Supplier No. HY10349A10MM 1ML
WAY-100635 maleate is a potent and selective 5-HT1A receptor antagonist used in pharmacological research. It is supplied as a high-purity solid or as a 10 mM solution in DMSO and is intended for in vitro and in vivo studies requiring serotonin 5-HT1A receptor blockade.
  • Potent, selective 5-HT1A receptor antagonist.
  • Available as 10 mM solution in DMSO or as solid powder.
  • High purity (≈99.6%) suitable for research use.
  • Recommended storage: solid at 4°C; solution at -20°C (short term) or -80°C (long term).