Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Way-100635 maleate | 1092679-51-0 | MFCD01321056 | 99.6% | 538.64 g/mol | C29H38N4O6 | 10 MG
SDP

Catalog No. 5000249307
Encompass_Preferred

WAY-100635 maleate is the maleate salt of WAY-100635, a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist supplied for research use in in vitro and in vivo pharmacology studies.

  • Potent 5-HT1A receptor antagonist (IC50 0.91 nM; Ki 0.39 nM)
  • High purity (≈99.6%) suitable for analytical and pharmacological studies
  • Molecular weight 538.64 g/mol; formula C29H38N4O6
  • Available in small pack sizes for research, including 10 mg
  • For research use only; not for human or veterinary use

Catalog No. 50-002-49307 Supplier Medchemexpress LLC Supplier No. HY10349A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WAY-100635 maleate is the maleate salt of WAY-100635, a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist supplied for research use in in vitro and in vivo pharmacology studies.

  • Potent 5-HT1A receptor antagonist (IC50 0.91 nM; Ki 0.39 nM)
  • High purity (≈99.6%) suitable for analytical and pharmacological studies
  • Molecular weight 538.64 g/mol; formula C29H38N4O6
  • Available in small pack sizes for research, including 10 mg
  • For research use only; not for human or veterinary use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.