Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Way-100635 maleate | 1092679-51-0 | MFCD01321056 | 99.6% | 538.64 g/mol | C29H38N4O6 | 100 MG
SDP

Catalog No. 5000248240
Encompass_Preferred

WAY-100635 maleate is the maleate salt of WAY-100635, a potent and selective 5-HT1A receptor antagonist used for pharmacology and receptor-binding research. It is provided as a high-purity analytical standard suitable for in vitro and in vivo studies.

  • Potent 5-HT1A receptor antagonist (IC50 = 0.91 nM; Ki = 0.39 nM).
  • High purity: 99.6% (HPLC).
  • Molecular weight 538.64 g/mol.
  • Molecular formula C29H38N4O6.
  • Supplied as a 100 mg vial for research use.

Catalog No. 50-002-48240 Supplier Medchemexpress LLC Supplier No. HY10349A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WAY-100635 maleate is the maleate salt of WAY-100635, a potent and selective 5-HT1A receptor antagonist used for pharmacology and receptor-binding research. It is provided as a high-purity analytical standard suitable for in vitro and in vivo studies.

  • Potent 5-HT1A receptor antagonist (IC50 = 0.91 nM; Ki = 0.39 nM).
  • High purity: 99.6% (HPLC).
  • Molecular weight 538.64 g/mol.
  • Molecular formula C29H38N4O6.
  • Supplied as a 100 mg vial for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.