Learn More
Medchemexpress LLC WAY-100635 maleate | 1092679-51-0 | MFCD01321056 | 99.6% | 538.64 g·mol⁻¹ | C29H38N4O6 | 50 MG
SDP
Catalog No. 5000246128
Encompass_Preferred
WAY-100635 maleate is the maleate salt of WAY-100635, a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist used in pharmacological research. It is supplied as a high-purity solid for use in receptor binding and functional assays.
  • Potent 5-HT1A antagonist with IC50 ~0.91 nM and Ki ~0.39 nM.
  • High purity, typically ≥99.5% by supplier analysis.
  • White to off-white solid suitable for formulation and assay work.
  • High solubility in DMSO; aqueous solutions may require sonication or warming.
  • Stable when sealed and stored cold; long-term solvent storage at -80°C is recommended.
Catalog No. 50-002-46128 Supplier Medchemexpress LLC Supplier No. HY10349A50MG
WAY-100635 maleate is the maleate salt of WAY-100635, a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist used in pharmacological research. It is supplied as a high-purity solid for use in receptor binding and functional assays.
  • Potent 5-HT1A antagonist with IC50 ~0.91 nM and Ki ~0.39 nM.
  • High purity, typically ≥99.5% by supplier analysis.
  • White to off-white solid suitable for formulation and assay work.
  • High solubility in DMSO; aqueous solutions may require sonication or warming.
  • Stable when sealed and stored cold; long-term solvent storage at -80°C is recommended.