Learn More
Selleck Chemical LLC WAY-100635 Maleate-10mg
SDP
Catalog No. 502390076
Encompass_Preferred
WAY-100635 Maleate is a potent and selective 5-HT receptor antagonist with IC50 of 0.95 nM.
Catalog No. 50-239-0076 Supplier Selleck Chemical LLC Supplier No. S266310MG
WAY-100635 Maleate is a potent and selective 5-HT receptor antagonist with IC50 of 0.95 nM.