Learn More
TARGETMOL CHEMICALS INC WAY-100635 maleate 25MG
SDP
Catalog No. 502878001
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. WAY-100635 Maleate is a specific and effective 5-HT receptor antagonist (IC50=0.95 nM). Purity 100%
Catalog No. 50-287-8001 Supplier TARGETMOL CHEMICALS INC Supplier No. T345325MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. WAY-100635 Maleate is a specific and effective 5-HT receptor antagonist (IC50=0.95 nM). Purity 100%