Learn More
Abcam WAY-100635 maleate, 5-HT1A receptor antagonist, 50MG
Catalog No. 502584585
Encompass_Preferred
MW 538.6 Da, Purity >98%. Potent, selective, silent 5-HT1A receptor antagonist (IC50 = 1.35 nM). Displays >100-fold selectivity for 5-HT1Arelative to a range of other CNS receptors including other 5-HT subtypes, adrenoceptors, dopamine, GABA, histamine and ion channels. Binds selectively to rodent brain 5-HT1A receptors *in vivo* (iv admin). Unlike WAY100135, WAY100635 displays no partial agonist activity in the ventral hippocampus of rats.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4585 Supplier Abcam Supplier No. AB12055050MG
MW 538.6 Da, Purity >98%. Potent, selective, silent 5-HT1A receptor antagonist (IC50 = 1.35 nM). Displays >100-fold selectivity for 5-HT1Arelative to a range of other CNS receptors including other 5-HT subtypes, adrenoceptors, dopamine, GABA, histamine and ion channels. Binds selectively to rodent brain 5-HT1A receptors *in vivo* (iv admin). Unlike WAY100135, WAY100635 displays no partial agonist activity in the ventral hippocampus of rats.
The product is subject to the following: Abcam Restricted Use Statement