Learn More
Medchemexpress LLC HY-10349A 5mg Medchemexpress, WAY-100635 Maleate CAS:1092679-51-0 Purity:>98%
SDP
Catalog No. 502080672
Encompass_Preferred
Medchemexpress, HY-10349A 5mg WAY-100635 Maleate CAS:1092679-51-0 WAY-100635 maleate is a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist with an IC50 value of 0.91 nM and Ki value of 0.39 nM. WAY-100635 maleate has pIC50 values for 5-HT1A and α1-adrenergic receptors of 8.9 and 6.6, respectively. WAY-100635 maleate is also a potent dopamine D4 receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0672 Supplier Medchemexpress LLC Supplier No. HY10349A5MG
Medchemexpress, HY-10349A 5mg WAY-100635 Maleate CAS:1092679-51-0 WAY-100635 maleate is a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist with an IC50 value of 0.91 nM and Ki value of 0.39 nM. WAY-100635 maleate has pIC50 values for 5-HT1A and α1-adrenergic receptors of 8.9 and 6.6, respectively. WAY-100635 maleate is also a potent dopamine D4 receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.