Learn More
Apexbio Technology LLC WAY-100635 maleate salt 1092679-51-0 100mg
SDP
Catalog No. 5000061557
Encompass_Preferred
WAY-100635 maleate salt (CAS 1092679-51-0) is a selective antagonist of the serotonin 5-HT1A receptor subtype It demonstrates high potency with a reported pIC50 value of 8 87 and displays over 100-fold selectivity relative to other serotonin receptor isoforms and common neurotransmitter receptor classes Due to its specific interaction with 5-HT1A receptors WAY-100635 maleate is frequently applied in neuroscience research to elucidate serotonin-mediated pathways and associated physiological or behavioral responses
Catalog No. 50-000-61557 Supplier Apexbio Technology LLC Supplier No. B2260100
WAY-100635 maleate salt (CAS 1092679-51-0) is a selective antagonist of the serotonin 5-HT1A receptor subtype It demonstrates high potency with a reported pIC50 value of 8 87 and displays over 100-fold selectivity relative to other serotonin receptor isoforms and common neurotransmitter receptor classes Due to its specific interaction with 5-HT1A receptors WAY-100635 maleate is frequently applied in neuroscience research to elucidate serotonin-mediated pathways and associated physiological or behavioral responses