Learn More
Cayman Chemical WAY-100635maleate 10mg
SDP
Catalog No. 502969971
Encompass_Preferred
A potent, silent antagonist of serotonin 5-HT1A receptors (IC50 = 2.2 nM; Ki = 0.8 nM) with 100-fold selectivity over other 5-HT subtypes; also exhibits agonist activity at dopamine D4 receptors (Kd = 2.4 nM)
Catalog No. 50-296-9971 Supplier Cayman Chemical Supplier No. 1459910 MG
A potent, silent antagonist of serotonin 5-HT1A receptors (IC50 = 2.2 nM; Ki = 0.8 nM) with 100-fold selectivity over other 5-HT subtypes; also exhibits agonist activity at dopamine D4 receptors (Kd = 2.4 nM)