Learn More
Enzo Life Sciences WAY-200070 (10mg). CAS: 440122-66-7
SDP
Catalog No. 502010952
Encompass_Preferred
Selective estrogen receptor ERβ agonist displaying 68-fold greater selectivity over ERα (EC50 = 2 and 155nM, respectively). It stimulates galanin expression in GnRH neurons and displays anxiolytic/antidepressant activity in various in vivo rodent models (administration to mice at 30mg/kg s.c.). Alternative name: 7-Bromo-2-(4-hydroxyphenyl)-1,3-benzoxazol-5-ol. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (12mg/ml) or 100% ethanol (2.5mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0952 Supplier Enzo Life Sciences Supplier No. BMLGR2470010
Selective estrogen receptor ERβ agonist displaying 68-fold greater selectivity over ERα (EC50 = 2 and 155nM, respectively). It stimulates galanin expression in GnRH neurons and displays anxiolytic/antidepressant activity in various in vivo rodent models (administration to mice at 30mg/kg s.c.). Alternative name: 7-Bromo-2-(4-hydroxyphenyl)-1,3-benzoxazol-5-ol. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (12mg/ml) or 100% ethanol (2.5mg/ml). Long Term Storage: Ambient.