Learn More
Cayman Chemical WAY-208466 (hydrochloride), Purity: ≥98%, CAS: 1207064-61-6, 10 mg
SDP
Catalog No. 502953879
Encompass_Preferred
A selective 5-HT6 receptor agonist (EC50 = 7.3 nM at the human 5-HT6 receptor); increases cortical GABA levels in rat frontal cortex, producing anxiolytic-like effects in vivo
Catalog No. 50-295-3879 Supplier Cayman Chemical Supplier No. 1193810 MG
A selective 5-HT6 receptor agonist (EC50 = 7.3 nM at the human 5-HT6 receptor); increases cortical GABA levels in rat frontal cortex, producing anxiolytic-like effects in vivo