Learn More
Medchemexpress LLC HY-107523A 5mg Medchemexpress, WAY-213613 (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 501968431
Encompass_Preferred
Medchemexpress, HY-107523A 5mg WAY-213613 (HCl) CAS: WAY-213613 hydrochloride is a potent, selective nonsubstrate reuptake inhibitor of GLT-1/EAAT2 with IC50 of 85 nM EAAT2. It displays 59- and 44-fold selectivity over EAAT1 and EAAT3 (IC50s are 5 and 3.8 μM, respectively). WAY-213613 hydrochloride shows no activity at ionotropic and metabotropic glutamate receptors. It is a potential tool for the elucidation of EAAT2 function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8431 Supplier Medchemexpress LLC Supplier No. HY107523A5MG
Medchemexpress, HY-107523A 5mg WAY-213613 (HCl) CAS: WAY-213613 hydrochloride is a potent, selective nonsubstrate reuptake inhibitor of GLT-1/EAAT2 with IC50 of 85 nM EAAT2. It displays 59- and 44-fold selectivity over EAAT1 and EAAT3 (IC50s are 5 and 3.8 μM, respectively). WAY-213613 hydrochloride shows no activity at ionotropic and metabotropic glutamate receptors. It is a potential tool for the elucidation of EAAT2 function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.