Learn More
Medchemexpress LLC Way-213613 hydrochloride | 2450268-84-3 | 99.8% | 451.65 g/mol | C16H14BrClF2N2O4 | 25 MG
SDP
Catalog No. 5000239768
Encompass_Preferred
WAY-213613 hydrochloride is a potent, selective inhibitor of the excitatory amino acid transporter 2 (EAAT2) used in biochemical and pharmacological research to study glutamate uptake and transporter-mediated signaling. The compound is supplied as a hydrochloride salt with well-defined chemical properties and high purity suitable for in vitro assays and mechanistic studies.
  • Potent and selective EAAT2 inhibition at nanomolar concentrations.
  • High purity appropriate for biochemical and cell-based assays.
  • Supplied as a hydrochloride salt to enhance stability and solubility.
  • Includes molecular weight and formula information for experimental planning.
  • Available in small-scale quantities for research workflows.
Catalog No. 50-002-39768 Supplier Medchemexpress LLC Supplier No. HY107523A25MG
WAY-213613 hydrochloride is a potent, selective inhibitor of the excitatory amino acid transporter 2 (EAAT2) used in biochemical and pharmacological research to study glutamate uptake and transporter-mediated signaling. The compound is supplied as a hydrochloride salt with well-defined chemical properties and high purity suitable for in vitro assays and mechanistic studies.
  • Potent and selective EAAT2 inhibition at nanomolar concentrations.
  • High purity appropriate for biochemical and cell-based assays.
  • Supplied as a hydrochloride salt to enhance stability and solubility.
  • Includes molecular weight and formula information for experimental planning.
  • Available in small-scale quantities for research workflows.