Learn More
Medchemexpress LLC Way-213613 hydrochloride | 2450268-84-3 | 99.8% | 451.65 g·mol⁻¹ | C16H14BrClF2N2O4 | 50 MG
SDP
Catalog No. 5000239798
Encompass_Preferred
WAY-213613 hydrochloride is the hydrochloride salt of WAY-213613, a potent and selective inhibitor of the human excitatory amino acid transporter 2 (EAAT2). It is supplied for research use in studies of glutamate transport and central nervous system pharmacology, and is reported to have an IC50 of 85 nM against human EAAT2.
  • Potent EAAT2 inhibition, IC50 85 nM.
  • High reported purity suitable for research use.
  • Available in multiple solid package sizes including 50 mg.
  • Suitable for in vitro and ex vivo glutamate transporter studies.
Catalog No. 50-002-39798 Supplier Medchemexpress LLC Supplier No. HY107523A50MG
WAY-213613 hydrochloride is the hydrochloride salt of WAY-213613, a potent and selective inhibitor of the human excitatory amino acid transporter 2 (EAAT2). It is supplied for research use in studies of glutamate transport and central nervous system pharmacology, and is reported to have an IC50 of 85 nM against human EAAT2.
  • Potent EAAT2 inhibition, IC50 85 nM.
  • High reported purity suitable for research use.
  • Available in multiple solid package sizes including 50 mg.
  • Suitable for in vitro and ex vivo glutamate transporter studies.