Learn More
Medchemexpress LLC HY-11035 50mg Medchemexpress, WAY-262611 CAS:1123231-07-1 Purity:>98%
SDP
Catalog No. 501968792
Encompass_Preferred
Medchemexpress, HY-11035 50mg WAY-262611 CAS:1123231-07-1 WAY-262611 is a wingless β-Catenin agonist that increases bone formation rate with an EC50 of 0.63 μM in TCF-Luciferase assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8792 Supplier Medchemexpress LLC Supplier No. HY1103550MG
Medchemexpress, HY-11035 50mg WAY-262611 CAS:1123231-07-1 WAY-262611 is a wingless β-Catenin agonist that increases bone formation rate with an EC50 of 0.63 μM in TCF-Luciferase assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.