Learn More
Abcam WAY 262611, Wnt pathway activator, 5MG
Catalog No. 502705379
Encompass_Preferred
MW 318.4 Da, Purity >97%. Wnt pathway activator. Selectively inhibits Dkk1 with no GSK-3β inhibition. TCF-Luciferase agonist (EC₅₀ = 0.63 μM). Increases bone formation rate in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5379 Supplier Abcam Supplier No. AB1452295MG
MW 318.4 Da, Purity >97%. Wnt pathway activator. Selectively inhibits Dkk1 with no GSK-3β inhibition. TCF-Luciferase agonist (EC₅₀ = 0.63 μM). Increases bone formation rate in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement